Anti-EXOSC6 Rabbit Polyclonal Antibody
Catalog # AVIVARP41087_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Exosome Component 6
- Antigen symbol:EXOSC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:118460
- Antigen synonyms:MTR3|p11|Mtr3p|EAP4|hMtr3p
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EXOSC6 (exosome component 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: