Anti-PCBP2 Rabbit Polyclonal Antibody
AVIVARP40567_T100
- Antibody type:Primary
- Antigen name:Poly(rC) binding protein 2
- Antigen symbol:PCBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5094
- Antigen synonyms:hnRNP-E2|HNRNPE2|HNRPE2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: PCBP2 (poly(rC) binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: