Anti-FBL Rabbit Polyclonal Antibody
Catalog # AVIVARP40366_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Fibrillarin
- Antigen symbol:FBL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2091
- Antigen synonyms:FIB|FLRN|RNU3IP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: