Order Entry
Slovenia
ContactUsLinkComponent
Anti-BRD3 Rabbit Polyclonal Antibody
Anti-BRD3 Rabbit Polyclonal Antibody
  AVIVARP38940_P050
undefined
Anti-BRD3 Rabbit Polyclonal Antibody
  AVIVARP38940_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    bromodomain containing 3
  • Antigen symbol:
    BRD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    8019
  • Antigen synonyms:
    ORFX|RING3L
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: BRD3 (bromodomain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: