Order Entry
Slovenia
ContactUsLinkComponent
Anti-STAT5B Rabbit Polyclonal Antibody
Anti-STAT5B Rabbit Polyclonal Antibody
  AVIVARP37146_T100
Anti-STAT5B Rabbit Polyclonal Antibody
  AVIVARP37146_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    Signal transducer and activator of transcription 5B
  • Antigen symbol:
    STAT5B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    20851
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: STAT5B (signal transducer and activator of transcription 5B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: