Anti-SCML1 Rabbit Polyclonal Antibody
Catalog # AVIVARP35812_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Sex Comb On Midleg-like 1
- Antigen symbol:SCML1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6322
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SCML1 (sex comb on midleg-like 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human