Anti-ASIC1 Rabbit Polyclonal Antibody
Catalog # AVIVARP35035_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Acid-sensing (proton-gated) ion channel 1
- Antigen symbol:ASIC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:11419
- Antigen synonyms:B530003N02Rik|BNaC2|Accn2|ASIC1a|AI843610|ASIC
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ASIC1 (acid-sensing (proton-gated) ion channel 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: