Order Entry
Slovenia
ContactUsLinkComponent
Anti-NGAL/Lipocalin 2 Rabbit Polyclonal Antibody
Anti-NGAL/Lipocalin 2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1373R
Supplier:  Bioss
undefined
Anti-NGAL/Lipocalin 2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1373R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lipocalin 2
  • Antigen symbol:
    NGAL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: NGAL/Lipocalin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat