Tag and ties holder
Catalog # BRDY800125
Supplier: Brady
Specifications
- Colour:Transparent
- Description:Tags/ties holder
- Material:Plastic
- W×D×H:114,3×38,1×190,5 mm
- Pk:1
Specifications
About this item
This wall-mounted holder is used to keep the tags, ties and marking pens to handy. It holds 50 tags, ties and marking pen and comes with adhesive strips and screws for mounting in a variety of ways.