Microbial air sampler, RCS® High Flow Touch
Catalog # BTSM1.44194.0001
Supplier: Merck
Specifications
- Description:Microbial air sampler
- Airflow:100 l/min
- Pk:1
Specifications
About this item
High flow touch sampler designed to ensure high quality, to maintain a safe work environment.
- Common sterilisation and disinfection methods
- Short sampling times as well as ease of handling
- Convenient programming with an easy touch
- User-defined options: Individual volumes, time delay, interval sampling
- Portable, battery driven and lightweight unit
Ordering information: Includes power supply, serial RS232 cable, USB adapter, RCS® management software, rotor, protection cap, carrying case, calibration certificate, quick start guide and user manual.