Order Entry
Slovenia
ContactUsLinkComponent
Accessories for bottle top vacuum filtration systems Nalgene®
Accessories for bottle top vacuum filtration systems Nalgene®
Catalog # 513-3971
   Low Manufacturing Impact
undefined
Accessories for bottle top vacuum filtration systems Nalgene®
Catalog # 513-3971
   Low Manufacturing Impact
Specifications
  • Description:
    Filter funnel adapter
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    25
Specifications
About this item
The non sterile PP adaptor is the bulk-packed version of funnel adapters included with 145- and 147-series analytical test filter funnels. Used in high throughput QC testing laboratories.