ESD safety shoes, lace-up, uvex 1, 8543
Supplier: UVEX
Certificates
About this item
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
- Climazone technology for improved comfort
- Breathable distance mesh lining
- Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
- Anti-slip sole with shock-absorbent heel and front of the foot
- Virtually seamless design eliminates potential pressure points and blisters
- Elastic laces for quick and individual adjustment
- 100% metal-free xenova® toecap, non magnetic (200 J)
- Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 11, 12
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC
Frequently Bought Together