Cap sealing mats for 96 microtubes
Supplier: MICRONIC
Certificates
About this item
The Micronic TPE push caps for tubes in 96-well format are compatible with the 0,50 ml, 0,75 ml, 1,10 ml, 1,40 ml, 2,00 ml, and 2,50 ml tubes. The full TPE cap range is available in natural or coloured for visual colour identification.
- Multi-pierceable without losing seal integrity
- High chemical resistance
- Sterile, gamma radiated or EtO treated
- RNase-, DNase- and pyrogen-free
- Manufactured in Class 7 cleanroom
Frequently Bought Together