Chemical resistant gloves, Showa 890
: Showa
These thin unsupported gloves with rolled cuffs are made of Viton®. They protect the hands from a wide array of chemical hazards (e.g. PCB and Hydrocarbon) and are therefore suitable for use in petrochemical industries.
- Impermeable for working in damp or greasy environments
- Cuff prevents dirt from entering the glove
- Wrist well protected
- Anatomical shape for perfect fit
Length: 350 mm
Thickness: 0,70 mm
Colour: Black
EN 388 (2101), EN 374 (DFL); Cat. III
: EN 388:2016 (1020X), EN ISO 374-1 Type B (DFL), EN ISO 374-5 VIRUS; Cat. III
- Catalog No:
- 112-5056
- 112-5055
- 112-5057
- 112-5058
- Model:
- Showa 890
- Showa 890
- Showa 890
- Showa 890
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 388:2016 (1020X), EN ISO 374-1 Type B (DFL), EN ISO 374-5 VIRUS
- EN 388:2016 (1020X), EN ISO 374-1 Type B (DFL), EN ISO 374-5 VIRUS
- EN 388:2016 (1020X), EN ISO 374-1 Type B (DFL), EN ISO 374-5 VIRUS
- EN 388:2016 (1020X), EN ISO 374-1 Type B (DFL), EN ISO 374-5 VIRUS
Frequently Bought Together