Order Entry
Slovenia
ContactUsLinkComponent
Screw caps with septa
Screw caps with septa
  SIAL27273
 :  Merck
Screw caps with septa
  SIAL27273
 :  Merck
  • For:
    2 ml large opening vials
  • Septum:
    PTFE/silicone
  • Description:
    Screw cap, black, PP, thread 10-425
  • Seq:
    01
  • Thickness:
    1,5 mm
  • Pk:
    1
Assembled PP screw caps with center hole and septa.