Leather gloves, Stronghand® Welderstar
These gloves are made of cowhide nappa leather, the cuff consists of split leather.
- Very soft and smooth leather
- Sewn with aramid fibres
Ideal for Argon welding applications.
Colour: Beige/grey
EN 388; DIN EN 12477 exposition A
: EN 388:2016 (3131X); DIN EN 12477 exposition A
- Catalog No:
- FELD0272/9
- FELD0272/8
- FELD0272/10
- FELD0272/11
- Model:
- Welderstar
- Welderstar
- Welderstar
- Welderstar
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (3131X), DIN EN 12477 exposition A
- EN 388:2016 (3131X), DIN EN 12477 exposition A
- EN 388:2016 (3131X), DIN EN 12477 exposition A
- EN 388:2016 (3131X), DIN EN 12477 exposition A
Frequently Bought Together