Hand disinfectants and cleansers with conditioner, Soft Care™ Sensisept H34
Catalog # DIVY6970900
Supplier: Diversey
Specifications
- Description:Hand disinfectant wash with conditioner, Soft Care™ Sensisept H34, 6x800 ml
- Seq:01
- Pk:6
Specifications
About this item
Soft Care™ Sensisept H34 is a highly effective disinfectant with superior mildness. It is a mild handwash product based on an surfactant system combined with chlorohexidine digluconate as a main bactericidal agent.
- Contains chlorhexidine digluconate, a highly effective bactericidal agent
- Slightly turbid and viscous liquid
- Relative density at 20 ºC: 1,04
- pH: 8,0
Caution: Use biocides safely. Always read the label and product information before use.