Order Entry
Slovenia
ContactUsLinkComponent
Surface disinfectant, SURFANIOS IP sterile
Surface disinfectant, SURFANIOS IP sterile
Catalog # SCUE871245
Supplier:  SCHUELKE MAYR
undefined
Surface disinfectant, SURFANIOS IP sterile
Catalog # SCUE871245
Supplier:  SCHUELKE MAYR
Specifications
  • Description:
    Surface disinfectant, SURFANIOS IP sterile, 15 ml single dose
  • Packed:
    Each carton contains 15 bags, each bag contains 8 ×15 ml single dose, plastic ampoules
  • Pk:
    1
Specifications
About this item
Sterile disinfectant for cleaning and disinfection of floors, surfaces, walls and equipment in pharmaceutical industry. Suitable for crictical areas and cleanroom classes A and B.
  • Bactericidal, fungicidal and virucidal
  • Contains N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, didecyldimethylammonium chloride, excipients.
  • Double bagged and irradiated
  • Aldehyde-free
Certifications: Bactericide EN 13697; Virucide 14476
Caution: Use biocides safely. Always read the label and product information before use.