Disinfectants, ClearKlens Cleansinald, sterile
Supplier: Diversey
Certificates
About this item
Surface active disinfectant based on a blend of quaternary ammonium and alkyl amines.
- 0,2 µm filtered, aseptically filled and double bagged
- Broad spectrum of activity
- Supplied with certificate of analysis and sterility where applicable
- Compatible with ClearKlens Tego® 2000, ClearKlens Tego® 2001 and ClearKlens Bi-Spore as a rotational process
Certifications: Passes CEN 1276, CEN 1650 and CEN 13697
Caution: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. Please contact your local Avantor office.
Articles may not be available in all countries. Please contact your local Avantor office.
Frequently Bought Together