Order Entry
Slovenia
ContactUsLinkComponent
Moisturising hand creams, Baktolan® cream
Moisturising hand creams, Baktolan® cream
Catalog # BODL972542
Supplier:  Bode
undefined
Moisturising hand creams, Baktolan® cream
Catalog # BODL972542
Supplier:  Bode

Specifications

  • Description:
    Moisturising hand cream, Baktolan® cream, 100 ml tube
  • Seq:
    01
  • Pk:
    100 mL

Specifications

About this item

Baktolan® cream is a replenishing and moisturising hand cream supporting the regeneration of highly stressed skin. It can be massaged into the dry skin several times a day.

  • W/O emulsion for dry and highly stressed skin
  • Increases the skin’s moisture content
  • Stabilises the skin’s protective acid mantle
  • Pleasant and mild scent
  • Free from preservatives
Caution: * These articles are only available in Germany.

*The product(s) on this page are outside the scope of the EU Directive 2019/904