Transport tubes, with stopper
Supplier: SEMADENI
Certificates
About this item
PS, clear, with white push-in stopper.
- Flat bottom
- Good thermal resistance
- Good chemical resistance
Universal tubes for a wide range of applications.
Frequently Bought Together