Chest wader, Steelite™ Safety S5, FW74
Supplier: Portwest
Certificates
About this item
Heavy-duty PVC/nitrile safety wader with adjustable straps, cord adjustable waist, stainless steel eyelets and integrated S5 safety boots.
- Colour-coded red outsole for easy identification of safety features
- Slip-and oil resistant, anti-static outsole
- Energy absorber heel
- Toe protection cap made from steel (200 J)
- Pierce resistant steel midsole
Colour: Black
EN ISO 20345:2011, S5
Certifications: EN ISO 20345:2011, S5
Specifications
- Catalog No:
- PRTWFW74BKR43
- PRTWFW74BKR39
- PRTWFW74BKR38
- PRTWFW74BKR45
- PRTWFW74BKR40
- PRTWFW74BKR41
- PRTWFW74BKR44
- PRTWFW74BKR42
- PRTWFW74BKR47
- PRTWFW74BKR46
- PRTWFW74BKR37
- Model:
- FW74
- FW74
- FW74
- FW74
- FW74
- FW74
- FW74
- FW74
- FW74
- FW74
- FW74
- Garment application:
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Oils, fats and fuels, Industrial
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Fabric:
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile
- Heavy-duty PVC/nitrile