Order Entry
Slovenia
ContactUsLinkComponent
Resealable Bags
Resealable Bags
Catalog # SEMA3431
Supplier:  SEMADENI
undefined
Resealable Bags
Catalog # SEMA3431
Supplier:  SEMADENI

Specifications

  • Description:
    Resealable bags with white strips, LDPE
  • L×W:
    90×75 mm
  • Seq:
    01
  • Thickness:
    0,05 mm
  • Pk:
    100

Specifications

About this item