ESD occupational footwear, clogs, ALPRO by Birkenstock®, C100
Supplier: LIEFERSERVICE UND LOGISTIK
Certificates
About this item
Breathable ESD clogs made from smooth leather with cork–Latex insole and anti-slip outer sole.
- Resistance range from 105 to 3,4×107 ohm
- Anti-slip, abrasion resistant sole (compliant with DIN 4843 part 100)
- Adjustable heel strap and instep strap
- Anatomical shape and deep heel cup
- Cushioning cork sole
For all areas where increased protection against electrostatic discharge is required.
Colour: White, blue or black
Certifications: EN ISO 61340
Frequently Bought Together