Cut resistant gloves, Tuff Cut Nit
Supplier: HONEYWELL SAFETY
Certificates
About this item
These gloves are knitted from Kevlar® yarn combined with stainless steel fibre. They feature a black nitrile foam coating on the palm.
- Stainless steel fibres provide maximum protection
- Long sleeve for increased protection of the wrist
- Nitrile coating provides good protection against oil and grease
- Foam coating is more flexible than standard nitrile coating
Ideal for handling heavy, very sharp parts with cutting edges, e.g. paper, metal and glass sheets and plastic parts. Also suitable for finishing operations with cutters.
Certifications: EN 388:2016 (4X43D), EN 407 (X1XXXX); Cat. II.
Specifications
- Catalog No:
- DALL2232533/10
- DALL2232533/8
- DALL2232533/9
- Model:
- Nit
- Nit
- Nit
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Kevlar®
- Kevlar®
- Kevlar®
- Finish:
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Gauge:
- 10
- 10
- 10
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)