Sumps for drums and small containers, PE-BS
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
These sumps of rotationally-molded body of black PE are used for storing water-endangering and aggressive media and suited for covering large-scale surfaces.
- Circumferential stabilising ribs
- Designed for placing flat on the floor
- Individual segments can be combined by galvanised element connectors
- Galvanised or PE grid support
- Maximum capacity 1000 kg/m2 with uniformly distributed load
Frequently Bought Together