SPE plates, 96-well, HyperSep™ Phenyl
Catalog # HYPE60300-688
Supplier: Thermo Fisher Scientific
Specifications
- Description:96-well plate, HyperSep™ Phenyl
- Seq:08
- Filling:100 mg
- Pk:1
Specifications
About this item
In Thermo Scientific™ HyperSep™ Phenyl Plates, the electron density of the aromatic ring provides different selectivity than other nonpolar sorbents for retention of basic compounds.
The cartridges are commonly used for nonpolar interactions, such as:
- Extraction of aromatic compounds
- Extraction of basic analytes