Order Entry
Slovenia
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) (from fish sperm), Sigma-Aldrich®
DNA (Deoxyribonucleic acid) (from fish sperm), Sigma-Aldrich®
Catalog # SIALD3159-10G
Supplier:  Merck
CAS Number:  
DNA (Deoxyribonucleic acid) (from fish sperm), Sigma-Aldrich®
Catalog # SIALD3159-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
EINECS:309-566-6
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922