Order Entry
Slovenia
Orders LinkContactUsLinkComponent
3,4-Dihydroxybenzoic acid

3,4-Dihydroxybenzoic acid

Supplier:   Cayman Chemical
3,4-Dihydroxybenzoic acid
Supplier:  Cayman Chemical
Certificates
3,4-Dihydroxybenzoic acid

3 Options Available Below

Product Details & Documents
Synonyms:  Protocatechuic acid
  • CAS Number:99-50-3
  • Formula:C₇H₆O₄
  • MDL Number:MFCD00002509
  • Molecular Weight:154.12 g/mol
  • Merck Index:12,08080
  • Boiling Pt:125…135 °C (0.001 mmHg)
  • Melting Pt:202…204 °C
  • Density:1.5415 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-760-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 63,00 €
    63,00 €
    1 * 25 g
       63,00 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 110,00 €
    110,00 €
    1 * 50 g
       110,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 195,00 €
    195,00 €
    1 * 100 g
       195,00 €