0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Trifluoroacetic acid
Trifluoroacetic acid 0.1% (v/v) in methanol
Trifluoroacetic acid 0.1% (v/v) in methanol
ROTHCP06.1
:
Roth Carl
:
Trifluoroacetic acid 0.1% (v/v) in methanol
ROTHCP06.1
:
Roth Carl
:
Pack type:
Glass bottle
Pk:
1 L
:
:
114.02 g/mol
:
Ambient
:
200-929-3
:
1230
:
3,II
:
76-05-1
:
C₂HF₃O₂
:
MFCD00004169