
Methyl-4-fluoro-3-nitrobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:329-59-9
:
Formula:C₈H₆FNO₄
MDL Number:MFCD00196161
:199.14 g/mol
Boiling Pt:109 °C (1 mmHg)
Melting Pt:56…59 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together