

Methyl trifluoromethanesulphonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:333-27-7
:
Formula:C₂H₃F₃O₃S
MDL Number:MFCD00000409
:164.11 g/mol
Boiling Pt:99 °C
Density:1.500
Flash Pt:38 °C (100 °F)
Storage Temperature:Refrigerator
EINECS:206-371-7
UN:2920
ADR:8,II
:GHS05
:GHS02
Frequently Bought Together