

(S)-(+)-Ethyl-3-piperidinecarboxylate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37675-18-6
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00792499
:157.21 g/mol
Density:1.043
Flash Pt:76 °C (169 °F)
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together