

4-Methyl-1,3-dioxane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1120-97-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00006569
Melting Pt:45…44 °C
Flash Pt:22 °C
Storage Temperature:Ambient
EINECS:214-323-1
UN:1993
ADR:3,II
:102.1317 g/mol
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4140 to 1.4160 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together