Order Entry
Slovenia
ContactUsLinkComponent
cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid, Sigma-Aldrich®
cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid, Sigma-Aldrich®
Catalog # SIALC112100-100G
Supplier:  Merck
CAS Number:  
cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid, Sigma-Aldrich®
Catalog # SIALC112100-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :246.17 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:223-256-7
CAS Number:3786-91-2
Formula:C₉H₁₀O₈
MDL Number:MFCD00001377