
4-Bromobenzenediazonium tetrafluoroborate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:673-40-5
:
Formula:C₆H₄BBrF₄N₂
MDL Number:MFCD00011894
:270.83 g/mol
Melting Pt:∼ 135 °C (dec.)
EINECS:211-608-2
ADR:8,III
:GHS05
Frequently Bought Together