Bathophenanthrolinedisulphonic acid disodium salt hydrate ≥95%, Sigma-Aldrich®
: 4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate
:52746-49-3
:53744-42-6
:C₂₄H₁₄N₂Na₂O₆S₂·H₂O
:MFCD00149310
:536.48 g/mol (anhy)
:300 °C (lit.)
:M-Clarity™ quality level = MQ200
:Room Temperature
:258-152-0
- Environmentally Preferable:
- Appearance (colour):White to Orange and Faint Beige to Beige and Pink
- Appearance (form):Powder
- Titration (NT) HCLO₄ 0.1M:calculated on dry substance
- Purity (HPLC area %):≥ 94.5 %
- Solubility (turbidity):Clear to Slightly Hazy
- Solubility (method):100 mg/ml in water
- Water:Report Result
Frequently Bought Together