Order Entry
Slovenia
ContactUsLinkComponent
1,5-Dimethyl-1H-pyrazol-3-carbaldehyde
1,5-Dimethyl-1H-pyrazol-3-carbaldehyde
Catalog # MAYBCC12604DA
Supplier:  Thermo Scientific
CAS Number:  
1,5-Dimethyl-1H-pyrazol-3-carbaldehyde
Catalog # MAYBCC12604DA
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :124.1405 g/mol
CAS Number:25016-10-8
Formula:C₆H₈N₂O
MDL Number:MFCD04967221