Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-DL-nipecotic acid ethyl ester
1-Boc-DL-nipecotic acid ethyl ester ≥97%
1-Boc-DL-nipecotic acid ethyl ester ≥97%
1-Boc-DL-nipecotic acid ethyl ester ≥97%
H32704.06
:
Thermo Fisher Scientific
:
1-Boc-DL-nipecotic acid ethyl ester ≥97%
H32704.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
:
130250-54-3
:
:
C₁₃H₂₃NO₄
:
MFCD04116274
:
257.33 g/mol
:
95 °C (0.5 mmHg)
:
31…35 °C
:
GHS06
:
GHS07
:
Ambient