Ionomycin calcium salt
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:56092-82-1
:
Formula:C₄₁H₇₀CaO₉
MDL Number:MFCD00151183
:747.1 g/mol
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
CHROMAN 1 (HYDROCHLORIDE HYDRATE) 1 * 1 mg
Supplier: Cayman Chemical