Safety storehouses for hazardous substances, SLH
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
The safety storehouse is a proved, mobile outdoor storage facility for hazardous substances.
- Base sump tray with grid support made from 3 mm fire galvanised sheet steel
- Reinforced roof and base profile frames
- Circumferential splash protection made from sheet steel, except in door area
- Galvanised steel double door with ventilation slots, 1950 mm wide, lockable with safety lock
- Built-in weather-protected ventilation slots in door and opposite rear panel
It can be transported unloaded at any time using a fork-lift truck or crane. This allows the system to be optimally adapted to suit the user’s actual requirements.
It is used for the storage of water endangering substances, active storage of flammable liquids a forced ventilation is required.
Certifications: Static approved, Z-38,5-171.
Frequently Bought Together