Storehouses for gas cylinders, GLH
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
These storehouses are ideal for outdoor storage of gas cylinders and gas cylinder pallets.
- Stable construction from special profiled galvanised sheet steel
- Galvanised double doors with cylinder lock
- Flexible holders and tension belts secure individual bottles against falling over
- High-loading galvanised grid-type floor also serves as ventilation underneath
These gas cylinder storehouses have been developed based on our widely used and practically proven safety storehouses. It provides closed construction and ideal protection against the weather and theft.
Certifications: Conforms with TRGS 510.
Frequently Bought Together