Safety cabinets for gas cylinders, SiZ G30
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
Safety cabinets are approved for storage of pressurised gas cylinders at the workplace.
- Stainless steel base plate- Niro Rhombus
- Leveling feet for installing cabinet horizontally
- Doors with cylinder lock
- Automatic thermal closing mechanism if fire occurs
Outer body made from powder-coated sheet steel, doors either in RAL 7035 - light grey or RAL 1007 - daffodil yellow and inner body made from HPL-coated sheets - RAL 7035.
Certifications: Conforms with DIN EN 14470-2 (G 30), DIN EN 14470-2, GS-approved safety (GS-sign), DIN 4102.
Delivery: Supplied with inner fittings: 1× stainless steel floor plate, 1× holder for 50 l gas cylinder per cylinder place, 2×horizontal C-rails for attaching gas fittings.
Frequently Bought Together