Storage cabinets for gas cylinders, GFL-F90
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
Gas cylinder storage system for regular storage outdoors of pressurised gas cylinders.
- Roof and walls in fire-resistant version
- Cantilever arm compartment divider with safety chains (fold out)
- Lockable grilled door
- Mounted delivery
- With hinged or double-hinged door depending on version
Certifications: Conforms with TRGS 510, DIN 4102.
Frequently Bought Together