Order Entry
Slovenia
ContactUsLinkComponent
VWR® Pre-mixed E. coli Media, for Biotechnology
VWR® Pre-mixed E. coli Media, for Biotechnology
Catalog # J910-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Pre-mixed E. coli Media, for Biotechnology
Catalog # J910-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 g
  • Description:
    M63 medium broth
  • Seq:
    02

Specifications

About this item

M9CA is a minimal medium supplemented with casamino acids used for the preparation of recombinant E. coli strains. M63 minimal medium is used for the cultivation and propagation of E. coli. Tryptone broth is used for the cultivation of E. coli. Also known as T-broth.

  • Save time by using pre-mixed, ready to use media
  • Homogenous, high quality powders

M9CA broth contains: Na₂HPO₄ (6 g/L), KH₂PO₄ (three g/L), NaCl (0,5 g/L), NH₄Cl (one g/L), casamino acids (two g/L) at a final pH 6,8 ±0,2 at 25 °C.
M63 medium broth contains: (NH₄)₂SO₄ (two g/L), KH₂PO₄ (13,6 g/L), FeSO₄.7H₂O (0,5 mg/L)
Tryptone broth contents: Tryptone (10 g/L), NaCl (five g/L) at a final pH 7,0 ±0,2 at 25 °C