Methyl 6-bromo-2-chloroquinoline-4-carboxylate
Catalog # MAYBRF00961.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:680213-43-8
Formula:C₁₁H₇BrClNO₂