4-Chloro-6-nitro-m-cresol, Sigma-Aldrich®
Catalog # SIAL146269-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:230-461-5
CAS Number:7147-89-9
Formula:C₇H₆ClNO₃
MDL Number:MFCD00007114