Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, 1,4-Dioxane 2000 µg/ml in dichloromethane
Organic reference standard, 1,4-Dioxane 2000 µg/ml in dichloromethane
Organic reference standard, 1,4-Dioxane 2000 µg/ml in dichloromethane
REST31853
:
Restek
:
Organic reference standard, 1,4-Dioxane 2000 µg/ml in dichloromethane
REST31853
:
Restek
:
Pack type:
Glass ampoule
Pk:
1 mL
CAS No.:
123-91-1
Description:
1,4-Dioxane 2,000 µg/ml in dichloromethane
Formula:
C₄H₈O₂
Description:
1,4-Dioxane