Diethylzinc 1 mol/l in hexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:557-20-0
:
Formula:C₄H₁₀Zn
MDL Number:MFCD00009021
:123.51 g/mol
:M-Clarity™ quality level = MQ100
EINECS:209-161-3
Frequently Bought Together