Industrial gloves, Titan 383
Supplier: MAPA
Certificates
About this item
These nitrile gloves with textile lining and smooth finish provide protection against humidity, oil and grease splashes.
- Good abrasion resistance
- Exceptionally comfortable lining: no seams on the working surfaces
- Last longer than precision work gloves, yet offers better comfort and dexterity than heavy-duty gloves
Ideal for precision mechanical work, handling oily mechanical parts, wood working, packaging and manufacturing small electrical components.
Certifications: EN 388:2016 (3111X), Cat. II
Specifications
- Catalog No:
- 112-5575
- 112-5576
- 112-5574
- Model:
- Titan 383
- Titan 383
- Titan 383
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Finish:
- Smooth
- Smooth
- Smooth
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3111X)
- EN 388:2016 (3111X)
- EN 388:2016 (3111X)